Learn More
MSC Mitutoyo Individual Rectangular Gage Block, 0.114 inch, Steel, Grade AS-1
Catalog No. 5000515973
Encompass
This precision gage block is an individual inch rectangular steel block, 0.114 inch in nominal length, designed for dimensional quality control. It offers high accuracy and is supplied with a Grade AS-1 accuracy certificate.
  • High accuracy, traceable to national standards.
  • Superior wringing performance due to advanced lapping.
  • Made from high-carbon, high-chrome steel for durability.
  • Heat-treated for hardness and long-term dimensional stability.
  • Provided with an inspection certificate.
  • Ideal for applications requiring repeated use of a single length.
Catalog No. 50-005-15973 Supplier MSC Supplier No. 88609110
This precision gage block is an individual inch rectangular steel block, 0.114 inch in nominal length, designed for dimensional quality control. It offers high accuracy and is supplied with a Grade AS-1 accuracy certificate.
  • High accuracy, traceable to national standards.
  • Superior wringing performance due to advanced lapping.
  • Made from high-carbon, high-chrome steel for durability.
  • Heat-treated for hardness and long-term dimensional stability.
  • Provided with an inspection certificate.
  • Ideal for applications requiring repeated use of a single length.