Learn More
MSC Mitutoyo Knife Straight Edge, Tool Steel, 8 in., 1 ea.
Catalog No. 5000516920
Encompass
This knife edge straight edge is designed for testing the straightness of workpiece features. It features a precision blade that provides accurate line contact with the work surface for reliable measurements.
  • Manufactured from tool steel
  • Features an acute angle of 30 degrees
  • Edge thickness of 0.004 in.
  • Blade is dyed black
  • Upper edge is covered with a grip plate
  • Supplied in a wooden case
  • Includes a protective cover
Catalog No. 50-005-16920 Supplier MSC Supplier No. 06252514
This knife edge straight edge is designed for testing the straightness of workpiece features. It features a precision blade that provides accurate line contact with the work surface for reliable measurements.
  • Manufactured from tool steel
  • Features an acute angle of 30 degrees
  • Edge thickness of 0.004 in.
  • Blade is dyed black
  • Upper edge is covered with a grip plate
  • Supplied in a wooden case
  • Includes a protective cover