Learn More
MSC Mitutoyo Limit Stickers Set, Green, 10 Pieces, For Series 2 Dial Indicator
Catalog No. 5000527949
Encompass
This Limit Stickers Set is designed for use with Series 2 Dial Indicators. It features a green color and includes 10 pieces per set, specifically for 55.6 mm or 57 mm bezel/outside diameter models to indicate tolerance markings.
  • Designed for Series 2 Dial Indicators
  • Features a vibrant green color
  • Includes 10 pieces per set
  • Compatible with 55.6 mm or 57 mm bezel/outside diameter models
  • Used for indicating tolerance markings
Catalog No. 50-005-27949 Supplier MSC Supplier No. 77036002
This Limit Stickers Set is designed for use with Series 2 Dial Indicators. It features a green color and includes 10 pieces per set, specifically for 55.6 mm or 57 mm bezel/outside diameter models to indicate tolerance markings.
  • Designed for Series 2 Dial Indicators
  • Features a vibrant green color
  • Includes 10 pieces per set
  • Compatible with 55.6 mm or 57 mm bezel/outside diameter models
  • Used for indicating tolerance markings