Learn More
MSC Mitutoyo Magnetic Indicator Back, 44 mm Diameter, 8 mm Thickness, AGD 3,4
Catalog No. 5000530476
Encompass
This product is a magnetic indicator back designed for specialized applications where the original indicator back may need to be replaced. It features a 44 mm diameter and 8 mm thickness, making it suitable for use with AGD 3,4 compatible indicators.
  • Magnetic back type
  • Designed for use with AGD 3,4 indicators
  • 44 mm diameter
  • 8 mm thickness
  • Ideal for special applications requiring indicator back replacement
Catalog No. 50-005-30476 Supplier MSC Supplier No. 90031170
This product is a magnetic indicator back designed for specialized applications where the original indicator back may need to be replaced. It features a 44 mm diameter and 8 mm thickness, making it suitable for use with AGD 3,4 compatible indicators.
  • Magnetic back type
  • Designed for use with AGD 3,4 indicators
  • 44 mm diameter
  • 8 mm thickness
  • Ideal for special applications requiring indicator back replacement