Learn More
MSC Mitutoyo Mechanical Indicating Micrometer, 1-2 inch, 0.0001 inch Graduation, IP54
Catalog No. 5000530551
Encompass
This mechanical indicating micrometer is designed for precise measurements in various applications, including three-wire measurements of pitch diameter for precision screws and parallelism measurements. It features a retractable anvil for faster workpiece throughput and carbide-tipped measuring faces for enhanced durability. The instrument is equipped with an IP54-rated housing, providing protection against dust and water, and comes with a fitted plastic case for convenient storage and transport.
  • Retractable anvil allows for faster workpiece throughput.
  • Carbide-tipped measuring faces resist wear.
  • IP54-rated housing provides protection against debris, dust, and water.
  • Ratchet-stop mechanism ensures uniform pressure for repeatable measurements.
  • Satin chrome finish resists rust and reduces glare.
  • Supplied with a fitted plastic case.
Catalog No. 50-005-30551 Supplier MSC Supplier No. 85798064
This mechanical indicating micrometer is designed for precise measurements in various applications, including three-wire measurements of pitch diameter for precision screws and parallelism measurements. It features a retractable anvil for faster workpiece throughput and carbide-tipped measuring faces for enhanced durability. The instrument is equipped with an IP54-rated housing, providing protection against dust and water, and comes with a fitted plastic case for convenient storage and transport.
  • Retractable anvil allows for faster workpiece throughput.
  • Carbide-tipped measuring faces resist wear.
  • IP54-rated housing provides protection against debris, dust, and water.
  • Ratchet-stop mechanism ensures uniform pressure for repeatable measurements.
  • Satin chrome finish resists rust and reduces glare.
  • Supplied with a fitted plastic case.