Learn More
MSC Mitutoyo Mechanical Inside Micrometer, 3.0000-3.5000 in Range, 0.000150 in Accuracy
Catalog No. 5000529716
Encompass
The Mitutoyo Mechanical Inside Micrometer is designed for precise internal diameter measurements. It features a highly stable three-point self-centering action, ensuring accurate readings closer to the bottom of blind bores. This durable instrument is suitable for a variety of laboratory and industrial applications.
  • Highly stable three-point self-centering action
  • 3 anvils evenly spaced at 120 degrees for accurate ID measurement
  • Measurement can be taken closer to the bottom of the blind bore
  • Can measure deep holes using an extension rod (optional)
  • Contact points and cone made from an alloy tool steel with a hardness of HRC60.5 or more
  • Micrometer-based movement
  • Ratchet stop thimble
  • Includes plastic case
Catalog No. 50-005-29716 Supplier MSC Supplier No. 06414635
The Mitutoyo Mechanical Inside Micrometer is designed for precise internal diameter measurements. It features a highly stable three-point self-centering action, ensuring accurate readings closer to the bottom of blind bores. This durable instrument is suitable for a variety of laboratory and industrial applications.
  • Highly stable three-point self-centering action
  • 3 anvils evenly spaced at 120 degrees for accurate ID measurement
  • Measurement can be taken closer to the bottom of the blind bore
  • Can measure deep holes using an extension rod (optional)
  • Contact points and cone made from an alloy tool steel with a hardness of HRC60.5 or more
  • Micrometer-based movement
  • Ratchet stop thimble
  • Includes plastic case