Learn More
MSC Mitutoyo Mechanical Outside Micrometer, 0-1 in, 0.0001 in Graduation, Friction Thimble
Catalog No. 5000529853
Encompass
The Mitutoyo Mechanical Outside Micrometer is designed for precise external measurements. It features a 0-1 inch range with a fine graduation of 0.0001 inch, a friction thimble for consistent pressure, and a mechanical digit counter for quick, error-free readings. Built for durability, it includes carbide-tipped measuring faces and a satin chrome finish, and comes with a fitted plastic case for protection.
  • Equipped with a mechanical digit counter for rapid and accurate readings up to 0.001 "
  • Obtains 0.0001 inch readings using its vernier scale.
  • Features carbide-tipped measuring faces for enhanced durability.
  • Has a satin chrome finish for corrosion resistance.
Catalog No. 50-005-29853 Supplier MSC Supplier No. 06243653
The Mitutoyo Mechanical Outside Micrometer is designed for precise external measurements. It features a 0-1 inch range with a fine graduation of 0.0001 inch, a friction thimble for consistent pressure, and a mechanical digit counter for quick, error-free readings. Built for durability, it includes carbide-tipped measuring faces and a satin chrome finish, and comes with a fitted plastic case for protection.
  • Equipped with a mechanical digit counter for rapid and accurate readings up to 0.001 "
  • Obtains 0.0001 inch readings using its vernier scale.
  • Features carbide-tipped measuring faces for enhanced durability.
  • Has a satin chrome finish for corrosion resistance.