Learn More
MSC Mitutoyo Mechanical Outside Micrometer Set, 13 Pc, 0 to 6 in, Ratchet Stop
Catalog No. 5000510376
Encompass
This mechanical outside micrometer set offers precise measurements for various applications. Designed with a ratchet stop thimble and solid carbide measuring faces, it ensures consistent and accurate readings. The 13-piece set includes six micrometers and five standards, covering a measurement range from 0 to 6 inches.
  • Minimum measurement of 0.0000 decimal inches.
  • Maximum measurement of 6.0000 decimal inches.
  • Features a ratchet stop thimble type for consistent pressure.
  • Equipped with solid carbide measuring faces for durability.
  • Finished with baked enamel for protection.
  • Includes a wrench, five standards, six micrometers, and a case.
  • Standards are included for 1, 2, 3, 4, and 5 inch measurements.
Catalog No. 50-005-10376 Supplier MSC Supplier No. 45869518
This mechanical outside micrometer set offers precise measurements for various applications. Designed with a ratchet stop thimble and solid carbide measuring faces, it ensures consistent and accurate readings. The 13-piece set includes six micrometers and five standards, covering a measurement range from 0 to 6 inches.
  • Minimum measurement of 0.0000 decimal inches.
  • Maximum measurement of 6.0000 decimal inches.
  • Features a ratchet stop thimble type for consistent pressure.
  • Equipped with solid carbide measuring faces for durability.
  • Finished with baked enamel for protection.
  • Includes a wrench, five standards, six micrometers, and a case.
  • Standards are included for 1, 2, 3, 4, and 5 inch measurements.