Learn More
MSC Mitutoyo Mechanical Outside Micrometer Set, 3 piece, 0 to 3 in, Friction & Ratchet Stop Thimble
Catalog No. 5000511187
Encompass
This Mechanical Outside Micrometer Set provides precise measurements from 0 to 3 inches. Designed for accuracy, it features a Friction & Ratchet Stop Thimble and Solid Carbide Measuring Face, ensuring reliable performance in various applications.
  • Minimum measurement is 0.0000 inch
  • Maximum measurement is 3.0000 inch
  • Accuracy is 0.000100 inch
  • Ratchet stop and friction thimble type
  • Solid carbide measuring face
  • Includes 3 pieces
  • Comes with a plastic case for storage
  • Manufactured in Japan
Catalog No. 50-005-11187 Supplier MSC Supplier No. 06243976
This Mechanical Outside Micrometer Set provides precise measurements from 0 to 3 inches. Designed for accuracy, it features a Friction & Ratchet Stop Thimble and Solid Carbide Measuring Face, ensuring reliable performance in various applications.
  • Minimum measurement is 0.0000 inch
  • Maximum measurement is 3.0000 inch
  • Accuracy is 0.000100 inch
  • Ratchet stop and friction thimble type
  • Solid carbide measuring face
  • Includes 3 pieces
  • Comes with a plastic case for storage
  • Manufactured in Japan