Learn More
MSC Mitutoyo Micrometer Anvil, 60 Degrees, High-Grade Steel, 24-14 Pitch
Catalog No. 5000533359
Encompass
This micrometer anvil is designed for use with 326 and 126 Series Screw Thread Micrometers. It features a 60 degrees anvil type and is constructed from high-grade steel, suitable for various measurement applications. The pitch ranges from 24 to 14.00.
  • Anvil type for screw thread micrometers
  • Compatible with 326 and 126 series screw thread micrometers
  • 60 degrees anvil type
  • Constructed from high-grade steel
  • Pitch range from 24 to 14.00
  • Manufactured in Japan
Catalog No. 50-005-33359 Supplier MSC Supplier No. 06234132
This micrometer anvil is designed for use with 326 and 126 Series Screw Thread Micrometers. It features a 60 degrees anvil type and is constructed from high-grade steel, suitable for various measurement applications. The pitch ranges from 24 to 14.00.
  • Anvil type for screw thread micrometers
  • Compatible with 326 and 126 series screw thread micrometers
  • 60 degrees anvil type
  • Constructed from high-grade steel
  • Pitch range from 24 to 14.00
  • Manufactured in Japan