Learn More
MSC Mitutoyo Micrometer Anvil, 60 Degrees, High-Grade Steel, for 326/126 Series
Catalog No. 5000512704
Encompass
This micrometer anvil is an accessory designed for use with 326 and 126 Series Screw Thread Micrometers. It features a 60-degree anvil made from high-grade steel, ensuring durability and precision for specialized measurement applications.
  • Accessory type: Anvil
  • For use with 326 and 126 Series Screw Thread Micrometers
  • 60-degree anvil type
  • Constructed from high-grade steel
  • Country of origin: Japan
Catalog No. 50-005-12704 Supplier MSC Supplier No. 06234090
This micrometer anvil is an accessory designed for use with 326 and 126 Series Screw Thread Micrometers. It features a 60-degree anvil made from high-grade steel, ensuring durability and precision for specialized measurement applications.
  • Accessory type: Anvil
  • For use with 326 and 126 Series Screw Thread Micrometers
  • 60-degree anvil type
  • Constructed from high-grade steel
  • Country of origin: Japan