Learn More
MSC Mitutoyo Micrometer Head, 0-0.5 inch Range, 0.001 inch Graduations, Spherical
Catalog No. 5000532953
Encompass
This micrometer head provides precise measurements with a 0-0.5 inch range and 0.001 inch graduations. It is equipped with a spherical spindle face for accurate readings and a zero-adjustable thimble for convenience, allowing the thimble to be set to zero at any position.
  • Thimble can be set to zero at any position.
  • Measuring range of 0-0.5 "
  • Graduations of 0.001 "
  • Accuracy of +/- 0.0001 "
  • Stem diameter is 0.375 "
  • Features a spherical spindle face.
  • Plain stem design.
  • Hard-chrome plating on scale surface.
Catalog No. 50-005-32953 Supplier MSC Supplier No. 06236194
This micrometer head provides precise measurements with a 0-0.5 inch range and 0.001 inch graduations. It is equipped with a spherical spindle face for accurate readings and a zero-adjustable thimble for convenience, allowing the thimble to be set to zero at any position.
  • Thimble can be set to zero at any position.
  • Measuring range of 0-0.5 "
  • Graduations of 0.001 "
  • Accuracy of +/- 0.0001 "
  • Stem diameter is 0.375 "
  • Features a spherical spindle face.
  • Plain stem design.
  • Hard-chrome plating on scale surface.