Learn More
MSC Mitutoyo Micrometer Head, 0-.5 in, 0.001 in Graduation, Flat Spindle, With Clamp Nut
Catalog No. 5000529133
Encompass
This Mitutoyo micrometer head is designed for precise measurements, featuring a 0-.5 in measuring range and a flat spindle end. It is suitable for various laboratory and industrial applications requiring accurate depth or dimension readings.
  • 0-.5 in measuring range
  • 0.001 in graduation
  • Flat spindle end
  • Stem with clamp nut
  • .375 in stem diameter
  • Measuring face made of alloy tool steel
  • Satin-chrome plated scale for clear readings
  • High accuracy of ±.0001 in
Catalog No. 50-005-29133 Supplier MSC Supplier No. 09529132
This Mitutoyo micrometer head is designed for precise measurements, featuring a 0-.5 in measuring range and a flat spindle end. It is suitable for various laboratory and industrial applications requiring accurate depth or dimension readings.
  • 0-.5 in measuring range
  • 0.001 in graduation
  • Flat spindle end
  • Stem with clamp nut
  • .375 in stem diameter
  • Measuring face made of alloy tool steel
  • Satin-chrome plated scale for clear readings
  • High accuracy of ±.0001 in