Learn More
MSC Mitutoyo Micrometer Standard, 16 in, ±0.00025 in Accuracy, 0.37 in Diameter
Catalog No. 5000529801
Encompass
The Mitutoyo micrometer standard is designed to precisely set the zero point for outside micrometers, ensuring accurate and repeatable measurements. These standards are manufactured with high precision, featuring exceptionally flat and lapped measuring faces. A heat-insulating handle helps prevent expansion from body temperature, contributing to consistent measurement accuracy.
  • Used for setting the zero point of outside micrometers.
  • Exceptional flatness and lapped measuring faces for precise measurements.
  • Heat insulating handle helps prevent expansion due to body temperature.
  • Available in inch and metric lengths.
  • Supplied in a fitted carton for standards up to 20 inch in length.
Catalog No. 50-005-29801 Supplier MSC Supplier No. 06238950
The Mitutoyo micrometer standard is designed to precisely set the zero point for outside micrometers, ensuring accurate and repeatable measurements. These standards are manufactured with high precision, featuring exceptionally flat and lapped measuring faces. A heat-insulating handle helps prevent expansion from body temperature, contributing to consistent measurement accuracy.
  • Used for setting the zero point of outside micrometers.
  • Exceptional flatness and lapped measuring faces for precise measurements.
  • Heat insulating handle helps prevent expansion due to body temperature.
  • Available in inch and metric lengths.
  • Supplied in a fitted carton for standards up to 20 inch in length.