Learn More
MSC Mitutoyo Outside Micrometer, 0.001 in Graduation, Ratchet Stop Thimble, Solid Carbide Anvil
Catalog No. 5000516670
Encompass
This mechanical outside micrometer provides precise measurements with a 0.001 in graduation. It features a ratchet stop thimble for consistent measuring force and solid carbide measuring faces for durability. The micrometer is designed with a heat insulating frame and includes a protective plastic case.
  • Precise 0.001 in graduation for accurate measurements.
  • Ratchet stop thimble ensures consistent measuring force.
  • Solid carbide anvil and measuring faces offer durability and wear resistance.
  • Spindle diameter of 6.35 mm.
  • Heat insulating frame for stable measurements.
  • Includes a protective plastic case for storage.
  • Proprietary inspection certification ensures quality.
  • Throat depth of 30 mm.
Catalog No. 50-005-16670 Supplier MSC Supplier No. 04081352
This mechanical outside micrometer provides precise measurements with a 0.001 in graduation. It features a ratchet stop thimble for consistent measuring force and solid carbide measuring faces for durability. The micrometer is designed with a heat insulating frame and includes a protective plastic case.
  • Precise 0.001 in graduation for accurate measurements.
  • Ratchet stop thimble ensures consistent measuring force.
  • Solid carbide anvil and measuring faces offer durability and wear resistance.
  • Spindle diameter of 6.35 mm.
  • Heat insulating frame for stable measurements.
  • Includes a protective plastic case for storage.
  • Proprietary inspection certification ensures quality.
  • Throat depth of 30 mm.