Learn More
MSC Mitutoyo Outside Micrometer, 0-25 mm, 0.01 mm Graduation, Ratchet Stop
Catalog No. 5000533281
Encompass
This outside micrometer is engineered for precise dimensional measurements in various industrial and laboratory settings. Its robust construction and reliable measuring mechanism ensure consistent and accurate readings.
  • Measuring range of 0-25 mm.
  • Graduation of 0.01 mm for detailed readings.
  • Ratchet stop for exact repetitive measurements.
  • Hammertone, baked-enamel-finished frame for enhanced durability.
  • Supplied in a fitted plastic case for secure storage and transport.
  • Carbide-tipped measuring faces for long-lasting wear resistance.
  • High accuracy of ±2μm for reliable results.
Catalog No. 50-005-33281 Supplier MSC Supplier No. 06230718
This outside micrometer is engineered for precise dimensional measurements in various industrial and laboratory settings. Its robust construction and reliable measuring mechanism ensure consistent and accurate readings.
  • Measuring range of 0-25 mm.
  • Graduation of 0.01 mm for detailed readings.
  • Ratchet stop for exact repetitive measurements.
  • Hammertone, baked-enamel-finished frame for enhanced durability.
  • Supplied in a fitted plastic case for secure storage and transport.
  • Carbide-tipped measuring faces for long-lasting wear resistance.
  • High accuracy of ±2μm for reliable results.