Learn More
MSC Mitutoyo Outside Micrometer, 50-75 mm, 0.01 mm Graduation, Ratchet Stop
Catalog No. 5000529680
Encompass
This outside micrometer features a measuring range of 50-75 mm and a graduation of 0.01 mm, designed for precise measurements. It includes a ratchet stop thimble for consistent, repetitive readings and carbide-tipped measuring faces for durability. The frame is finished with a hammertone, baked-enamel coating.
  • Hammertone, baked-enamel-finished frame.
  • Ratchet stop for exact repetitive readings.
  • Includes a standard bar.
  • Carbide-tipped measuring faces.
  • Lightweight workshop design.
  • Provides accurate measurements with ±2μm accuracy.
Catalog No. 50-005-29680 Supplier MSC Supplier No. 06230759
This outside micrometer features a measuring range of 50-75 mm and a graduation of 0.01 mm, designed for precise measurements. It includes a ratchet stop thimble for consistent, repetitive readings and carbide-tipped measuring faces for durability. The frame is finished with a hammertone, baked-enamel coating.
  • Hammertone, baked-enamel-finished frame.
  • Ratchet stop for exact repetitive readings.
  • Includes a standard bar.
  • Carbide-tipped measuring faces.
  • Lightweight workshop design.
  • Provides accurate measurements with ±2μm accuracy.