Learn More
MSC Mitutoyo Point Micrometer, 0-25 mm, 0.01 mm Graduation, 30 Degree Point
Catalog No. 5000528198
Encompass
This point micrometer features pointed tips on the anvil and spindle, making it ideal for measuring small grooves, keyways, and other hard-to-reach areas. It offers precise measurements for various workpieces.
  • Ratchet mechanism ensures consistent measurement force.
  • Vernier scale graduated to 0.01 mm for precise outer dimension measurements.
  • Spindle lock provides secure locking of the measurement position.
  • Satin chrome finish and laser etched measurements for easier reading.
  • Frame constructed of high-grade steel with a baked enamel finish for durability.
  • Supplied in a fitted plastic case for protection.
  • Points are constructed of hardened steel.
Catalog No. 50-005-28198 Supplier MSC Supplier No. 35450956
This point micrometer features pointed tips on the anvil and spindle, making it ideal for measuring small grooves, keyways, and other hard-to-reach areas. It offers precise measurements for various workpieces.
  • Ratchet mechanism ensures consistent measurement force.
  • Vernier scale graduated to 0.01 mm for precise outer dimension measurements.
  • Spindle lock provides secure locking of the measurement position.
  • Satin chrome finish and laser etched measurements for easier reading.
  • Frame constructed of high-grade steel with a baked enamel finish for durability.
  • Supplied in a fitted plastic case for protection.
  • Points are constructed of hardened steel.