Learn More
MSC Mitutoyo Rectangle Steel Gage Block, 1 in, Grade 0
Catalog No. 5000533166
Encompass
This precision gage block is designed for accurate measurement applications. Made from high-quality steel, it features a rectangular shape and an accuracy grade of 0, ensuring high precision and reliability. Each block is supplied with NIST certification, providing traceable calibration for quality assurance.
  • Rectangle shape
  • Grade 0 accuracy
  • Constructed from steel
  • 1 inch length
  • Includes NIST certification
Catalog No. 50-005-33166 Supplier MSC Supplier No. 14344386
This precision gage block is designed for accurate measurement applications. Made from high-quality steel, it features a rectangular shape and an accuracy grade of 0, ensuring high precision and reliability. Each block is supplied with NIST certification, providing traceable calibration for quality assurance.
  • Rectangle shape
  • Grade 0 accuracy
  • Constructed from steel
  • 1 inch length
  • Includes NIST certification