Learn More
MSC Mitutoyo Rectangle Steel Gage Block, 5 mm, Grade 0
Catalog No. 5000515889
Encompass
This precision gage block is crafted from steel, featuring a rectangular shape and an overall length of 5 mm. It is engineered for high accuracy applications, meeting Grade 0 specifications.
  • Includes certificate of inspection
  • Manufactured to ASME standards
Catalog No. 50-005-15889 Supplier MSC Supplier No. 85389476
This precision gage block is crafted from steel, featuring a rectangular shape and an overall length of 5 mm. It is engineered for high accuracy applications, meeting Grade 0 specifications.
  • Includes certificate of inspection
  • Manufactured to ASME standards