Learn More
MSC Mitutoyo Rectangle Steel Gage Block, Steel, 0.119 in, Grade AS-1
Catalog No. 5000529583
Encompass
This precision rectangle gage block is made from steel and designed for highly accurate measurement applications. It features an AS-1 accuracy grade, ensuring reliable and consistent performance in demanding environments.
  • Includes certificate of inspection
  • Rectangular shape
  • Manufactured to AS-1 accuracy grade
  • Made from durable steel
  • Originates from Japan
Catalog No. 50-005-29583 Supplier MSC Supplier No. 01135235
This precision rectangle gage block is made from steel and designed for highly accurate measurement applications. It features an AS-1 accuracy grade, ensuring reliable and consistent performance in demanding environments.
  • Includes certificate of inspection
  • Rectangular shape
  • Manufactured to AS-1 accuracy grade
  • Made from durable steel
  • Originates from Japan