Learn More
MSC Mitutoyo Rectangular Steel Gage Block, 0.123 in, Grade AS-1
Catalog No. 5000533409
Encompass
This individual rectangular steel gage block is designed for precision measurements and is suitable for setting and calibrating fixtures and precision instruments. It is ideal for applications requiring the repeated use of a single length.
  • Length of 0.123 in.
  • Manufactured from steel for durability.
  • Accuracy Grade AS-1, intended for shop-floor use.
  • Supplied with an Inspection Certificate.
Catalog No. 50-005-33409 Supplier MSC Supplier No. 00396259
This individual rectangular steel gage block is designed for precision measurements and is suitable for setting and calibrating fixtures and precision instruments. It is ideal for applications requiring the repeated use of a single length.
  • Length of 0.123 in.
  • Manufactured from steel for durability.
  • Accuracy Grade AS-1, intended for shop-floor use.
  • Supplied with an Inspection Certificate.