Learn More
MSC Mitutoyo Replacement Head Assembly, 2.0-2.5 in, TiN-Coated
Catalog No. 5000513778
Encompass
This replacement head assembly is designed for use with Mitutoyo Holtest, Digimatic Holtest, and Borematic bore gages, offering a measuring range of 2.0-2.5 inches. It features TiN-coated measuring contacts for enhanced durability and precision. An optional adapter is needed to connect it to Borematic main body units.
  • TiN-coated measuring contact for excellent durability and impact resistance.
  • Enables measurements closer to the bottom of blind bore holes.
  • Compatible with Holtest Type 1, Digimatic Holtest, and Borematic systems.
Catalog No. 50-005-13778 Supplier MSC Supplier No. 70419080
This replacement head assembly is designed for use with Mitutoyo Holtest, Digimatic Holtest, and Borematic bore gages, offering a measuring range of 2.0-2.5 inches. It features TiN-coated measuring contacts for enhanced durability and precision. An optional adapter is needed to connect it to Borematic main body units.
  • TiN-coated measuring contact for excellent durability and impact resistance.
  • Enables measurements closer to the bottom of blind bore holes.
  • Compatible with Holtest Type 1, Digimatic Holtest, and Borematic systems.