Learn More
MSC Mitutoyo RS-Link Connecting Cable, 0.5m, for EH/EV Counters
Catalog No. 5000536125
Encompass
This connecting cable is designed to facilitate the RS Link Function, enabling multipoint measurement capabilities. It is used to connect EH/EV counters and RS units, allowing for versatile data acquisition in various measurement setups.
  • Connects EH/EV counters and RS units.
  • Supports multipoint measurement of up to 60 channels.
  • Allows up to 10 EV-16A units to be linked together.
  • Total length of all linked RS-link connecting cables limited to 10m.
  • Optional accessory for Mu-Checker Counter EV-16A.
  • Required for connecting Display Unit D-EV with an EV-16A.
Catalog No. 50-005-36125 Supplier MSC Supplier No. 76145671
This connecting cable is designed to facilitate the RS Link Function, enabling multipoint measurement capabilities. It is used to connect EH/EV counters and RS units, allowing for versatile data acquisition in various measurement setups.
  • Connects EH/EV counters and RS units.
  • Supports multipoint measurement of up to 60 channels.
  • Allows up to 10 EV-16A units to be linked together.
  • Total length of all linked RS-link connecting cables limited to 10m.
  • Optional accessory for Mu-Checker Counter EV-16A.
  • Required for connecting Display Unit D-EV with an EV-16A.