Learn More
MSC Mitutoyo Screw Checker, Acme Thread, 1-10 TPI/mm
Catalog No. 5000511691
Encompass
This screw checker is designed for accurately identifying Acme thread types and sizes. It allows for quick measurement of threads per inch and screw size within a specified range, making it a useful tool for various applications.
  • Identifies Acme thread types
  • Measures threads per inch from 1 to 10
  • Checks screw sizes from 2 to 10
  • Suitable for both inch and millimeter measurements
Catalog No. 50-005-11691 Supplier MSC Supplier No. 59372854
This screw checker is designed for accurately identifying Acme thread types and sizes. It allows for quick measurement of threads per inch and screw size within a specified range, making it a useful tool for various applications.
  • Identifies Acme thread types
  • Measures threads per inch from 1 to 10
  • Checks screw sizes from 2 to 10
  • Suitable for both inch and millimeter measurements