Learn More
MSC Mitutoyo Screw Pitch Gage, Metric, 28 Leaves, 0.25-2.5 mm, 60 Degree
Catalog No. 5000528497
Encompass
This screw pitch gage is designed for precise measurement of screw threads. It features 28 leaves, allowing for metric measurements across a pitch range of 0.25 to 2.5 mm and a 60-degree thread angle. Constructed from stainless steel, it is suitable for both internal and external thread locations and includes a locking mechanism for secure readings.
  • Measures metric threads
  • Features 28 leaves for various pitches
  • Covers a pitch range of 0.25 to 2.5 mm
  • Designed with a 60 degree thread angle
  • Suitable for internal and external thread measurement
  • Constructed from stainless steel for durability
  • Includes a locking mechanism for stability during measurement
Catalog No. 50-005-28497 Supplier MSC Supplier No. 47857438
This screw pitch gage is designed for precise measurement of screw threads. It features 28 leaves, allowing for metric measurements across a pitch range of 0.25 to 2.5 mm and a 60-degree thread angle. Constructed from stainless steel, it is suitable for both internal and external thread locations and includes a locking mechanism for secure readings.
  • Measures metric threads
  • Features 28 leaves for various pitches
  • Covers a pitch range of 0.25 to 2.5 mm
  • Designed with a 60 degree thread angle
  • Suitable for internal and external thread measurement
  • Constructed from stainless steel for durability
  • Includes a locking mechanism for stability during measurement