Learn More
MSC Mitutoyo Screw Thread Micrometer, Mechanical, 2 to 3 in, 0.001 in Graduation
Catalog No. 5000529729
Encompass
This precision measuring instrument is designed for screw thread measurements, featuring a ratchet stop thimble for consistent pressure. It offers a graduation of 0.001 in and an accuracy of 0.0002 in. The micrometer comes with interchangeable anvils and is housed in a plastic case. It has an Ingress Protection Rating of IP65 and is TUV certified.
  • Ratchet stop thimble for consistent pressure
  • 0.001 in graduation
  • 0.0002 in accuracy
  • Interchangeable anvils
  • Includes plastic case
  • Ingress Protection Rating IP65
  • TUV certified
Catalog No. 50-005-29729 Supplier MSC Supplier No. 06234256
This precision measuring instrument is designed for screw thread measurements, featuring a ratchet stop thimble for consistent pressure. It offers a graduation of 0.001 in and an accuracy of 0.0002 in. The micrometer comes with interchangeable anvils and is housed in a plastic case. It has an Ingress Protection Rating of IP65 and is TUV certified.
  • Ratchet stop thimble for consistent pressure
  • 0.001 in graduation
  • 0.0002 in accuracy
  • Interchangeable anvils
  • Includes plastic case
  • Ingress Protection Rating IP65
  • TUV certified