Learn More
MSC Mitutoyo Setting Ring, 0.5000 in ID, 1.2600 in OD, 0.000060 in Accuracy
Catalog No. 5000530297
Encompass
This setting ring is designed for precise calibration and measurement applications. It features specific inside and outside diameters with a high level of accuracy, making it suitable for demanding industrial and laboratory environments.
  • Inside diameter of 0.5000 in
  • Outside diameter of 1.2600 in
  • Accuracy of 0.000060 in
  • Manufactured in Japan
Catalog No. 50-005-30297 Supplier MSC Supplier No. 77614980
This setting ring is designed for precise calibration and measurement applications. It features specific inside and outside diameters with a high level of accuracy, making it suitable for demanding industrial and laboratory environments.
  • Inside diameter of 0.5000 in
  • Outside diameter of 1.2600 in
  • Accuracy of 0.000060 in
  • Manufactured in Japan