Learn More
MSC Mitutoyo Setting Ring, Ceramic, 16 mm Nominal Size, 45 mm OD
Catalog No. 5000533430
Encompass
This setting ring is designed for the quick and accurate setting of dial bore gages, Holtest, and inside micrometers.
  • Actual calibration value for the diameter is marked on each ring.
  • No anticorrosion treatment is required for ceramic setting rings, resulting in simple maintenance and storage.
  • Can be used for calibration when an optimal size is prepared.
Catalog No. 50-005-33430 Supplier MSC Supplier No. 06239370
This setting ring is designed for the quick and accurate setting of dial bore gages, Holtest, and inside micrometers.
  • Actual calibration value for the diameter is marked on each ring.
  • No anticorrosion treatment is required for ceramic setting rings, resulting in simple maintenance and storage.
  • Can be used for calibration when an optimal size is prepared.