Learn More
MSC Mitutoyo Setting Ring Gage, Metric, 200 mm, Steel
Catalog No. 5000528118
Encompass
This setting ring gage is a precision instrument designed for calibrating bore gages and micrometers. It features a 200 mm diameter and is constructed from durable steel, ensuring accurate metric measurements.
  • Can be used for calibration.
  • Actual calibration value for the diameter is marked on each ring.
Catalog No. 50-005-28118 Supplier MSC Supplier No. 08667768
This setting ring gage is a precision instrument designed for calibrating bore gages and micrometers. It features a 200 mm diameter and is constructed from durable steel, ensuring accurate metric measurements.
  • Can be used for calibration.
  • Actual calibration value for the diameter is marked on each ring.