Learn More
MSC Mitutoyo SPC Cable, For Digimatic Calipers, 2.00 m
Catalog No. 5000535440
Encompass
This SPC cable provides a direct data connection for Digimatic Calipers, enabling efficient data transfer for measurement applications.
  • Designed for use with Digimatic Calipers
  • Ensures reliable data communication
  • Provides a cable length of 2.00 meters
Catalog No. 50-005-35440 Supplier MSC Supplier No. 39888748
This SPC cable provides a direct data connection for Digimatic Calipers, enabling efficient data transfer for measurement applications.
  • Designed for use with Digimatic Calipers
  • Ensures reliable data communication
  • Provides a cable length of 2.00 meters