Learn More
MSC Mitutoyo SPC Cable, For Digimatic Scale Units, 40 in
Catalog No. 5000528412
Encompass
The Mitutoyo SPC Cable is designed for connecting Absolute Digimatic Scale Units and DP-1VR Series 264 Digimatic Mini-Processors. This cable ensures reliable data transfer for precise measurements.
  • Connects to Absolute Digimatic Scale Units
  • Connects to DP-1VR Series 264 Digimatic Mini-Processors
  • Compatible with calipers
  • Measures 40 in (1016 mm) in length
  • Manufactured in Japan
Catalog No. 50-005-28412 Supplier MSC Supplier No. 32155095
The Mitutoyo SPC Cable is designed for connecting Absolute Digimatic Scale Units and DP-1VR Series 264 Digimatic Mini-Processors. This cable ensures reliable data transfer for precise measurements.
  • Connects to Absolute Digimatic Scale Units
  • Connects to DP-1VR Series 264 Digimatic Mini-Processors
  • Compatible with calipers
  • Measures 40 in (1016 mm) in length
  • Manufactured in Japan