This is a mechanical spherical face micrometer designed for precise measurements up to 25mm. It features a ball anvil and ratchet thimble for consistent results, making it suitable for various laboratory and industrial applications requiring accurate dimensional checks.
- Mechanical operation for reliable performance.
- Ball anvil for specialized measurement tasks.
- Ratchet thimble ensures constant measuring force.
- Accuracy of ±3 μm for high precision.
- Graduation of 0.01 mm for fine readings.
- Constructed with durable steel anvil and spindle materials.
- Satin chrome finish for durability and readability.
- Includes a plastic case for protection and storage.
- IP65 ingress protection rating for resistance against dust and water.
- TUV Certified, ensuring compliance with quality standards.
- Powered by one SR44 battery (included).