Learn More
MSC Mitutoyo Square Steel Gage Block, 0.114 in, Grade 0, ASME
Catalog No. 5000531602
Encompass
This gage block is made of steel and designed for high-precision measurement. It features a square shape, a size of 0.114 inches, and an accuracy grade of 0, ensuring reliable performance for calibration and inspection tasks. Conforming to ASME standards, it comes with a Certificate of Inspection.
  • Accuracy grade 0
  • Includes certificate of inspection
  • Square shape
  • Conforms to ASME standards
  • Steel material
  • Size of 0.114 inches
  • Manufactured in Japan
Catalog No. 50-005-31602 Supplier MSC Supplier No. 88609565
This gage block is made of steel and designed for high-precision measurement. It features a square shape, a size of 0.114 inches, and an accuracy grade of 0, ensuring reliable performance for calibration and inspection tasks. Conforming to ASME standards, it comes with a Certificate of Inspection.
  • Accuracy grade 0
  • Includes certificate of inspection
  • Square shape
  • Conforms to ASME standards
  • Steel material
  • Size of 0.114 inches
  • Manufactured in Japan