Learn More
MSC Mitutoyo Square Steel Gage Block, 0.139 in, Grade 0
Catalog No. 5000512899
Encompass
This precision square steel gage block is a primary standard for dimensional quality control. Manufactured from high-carbon, high-chrome steel, it offers high accuracy and durability with excellent dimensional stability, minimizing deterioration over time. It meets or exceeds ASME B89.1.9-2002 specifications.
  • Secure wringing allows for perfect wringing due to a central hole.
  • Easily assemble a precision height reference standard using accessories.
  • Create dedicated, inexpensive inspection jigs for instrument inspection.
  • Wide measuring surface (24.1 x 24.1 mm) maintains stable orientation for diverse applications.
  • Guaranteed high accuracy, backed by a traceable system.
  • Includes certificate of inspection.
  • Suitable for inspection areas to verify accuracy of plug and snap gages.
Catalog No. 50-005-12899 Supplier MSC Supplier No. 08759946
This precision square steel gage block is a primary standard for dimensional quality control. Manufactured from high-carbon, high-chrome steel, it offers high accuracy and durability with excellent dimensional stability, minimizing deterioration over time. It meets or exceeds ASME B89.1.9-2002 specifications.
  • Secure wringing allows for perfect wringing due to a central hole.
  • Easily assemble a precision height reference standard using accessories.
  • Create dedicated, inexpensive inspection jigs for instrument inspection.
  • Wide measuring surface (24.1 x 24.1 mm) maintains stable orientation for diverse applications.
  • Guaranteed high accuracy, backed by a traceable system.
  • Includes certificate of inspection.
  • Suitable for inspection areas to verify accuracy of plug and snap gages.