Learn More
MSC Mitutoyo Surface Roughness Gage Stand, For SJ 210/310/410
Catalog No. 5000512931
Encompass
This surface roughness gage stand is designed for use with SJ 210/310/410 models of SJ surface roughness testers. It provides a stable base for accurate measurements in various applications.
  • Accessory type: Surface roughness gage stand
  • Designed for use with SJ 210/310/410 instrument types
  • Compatible with SJ surface roughness tester units
  • Overall depth: 7.0000 in
  • Overall length: 7.0000 in
Catalog No. 50-005-12931 Supplier MSC Supplier No. 77614659
This surface roughness gage stand is designed for use with SJ 210/310/410 models of SJ surface roughness testers. It provides a stable base for accurate measurements in various applications.
  • Accessory type: Surface roughness gage stand
  • Designed for use with SJ 210/310/410 instrument types
  • Compatible with SJ surface roughness tester units
  • Overall depth: 7.0000 in
  • Overall length: 7.0000 in