Learn More
MSC Mitutoyo Height Gauge Adapter, 9 mm x 9 mm, For Surftest, 1 ea
Catalog No. 5000511122
Encompass
This height gauge adapter allows for the seamless mounting of surface roughness testers to height gauges. It is specifically designed to expand the utility of equipment by providing a secure interface for components such as indicators, stylus tips, or scribers.
  • Compatible with Surftest portable surface roughness testers, including SJ-210, SJ-301, and SJ-310 models
  • Features 9 mm x 9 mm dimensions
  • Facilitates precise mounting of additional measurement accessories to standard height gauges
Catalog No. 50-005-11122 Supplier MSC Supplier No. 03300480
This height gauge adapter allows for the seamless mounting of surface roughness testers to height gauges. It is specifically designed to expand the utility of equipment by providing a secure interface for components such as indicators, stylus tips, or scribers.
  • Compatible with Surftest portable surface roughness testers, including SJ-210, SJ-301, and SJ-310 models
  • Features 9 mm x 9 mm dimensions
  • Facilitates precise mounting of additional measurement accessories to standard height gauges