Learn More
MSC Mitutoyo Telescoping Gage, 3/4 to 1-1/4 in., Satin Chrome Finish, 1 ea.
Catalog No. 5000512394
Encompass
This telescoping gage is designed for precise measurement of internal diameters, slots, and grooves. It is intended for use in conjunction with an outside micrometer to facilitate the accurate determination of bore or opening dimensions.
  • Features a spring-loaded plunger that expands within the bore or groove
  • Constructed with fully hardened steel gaging surfaces for extended tool life
  • Equipped with a knurled handle and clamp
  • Finished with a satin chrome coating
  • Measures between 3/4 in. to 1-1/4 in.
  • Overall length is 4.3307 in.
Catalog No. 50-005-12394 Supplier MSC Supplier No. 06237515
This telescoping gage is designed for precise measurement of internal diameters, slots, and grooves. It is intended for use in conjunction with an outside micrometer to facilitate the accurate determination of bore or opening dimensions.
  • Features a spring-loaded plunger that expands within the bore or groove
  • Constructed with fully hardened steel gaging surfaces for extended tool life
  • Equipped with a knurled handle and clamp
  • Finished with a satin chrome coating
  • Measures between 3/4 in. to 1-1/4 in.
  • Overall length is 4.3307 in.