Learn More
MSC Mitutoyo Telescoping Gage Set, 6 Piece, 8-150 mm Range, Steel
Catalog No. 5000529229
Encompass
This telescoping gage set is designed for precise inside diameter measurement. It is a 6-piece set, featuring rounded tips that compensate for the radius to ensure accurate readings.
  • Designed for inside diameter measurement.
  • Includes 6 telescoping gages.
  • Features rounded tips to compensate for the radius.
  • Measures diameters from 8 mm to 150 mm.
  • Constructed from durable steel.
  • Minimum overall length is 1 "
Catalog No. 50-005-29229 Supplier MSC Supplier No. 09503871
This telescoping gage set is designed for precise inside diameter measurement. It is a 6-piece set, featuring rounded tips that compensate for the radius to ensure accurate readings.
  • Designed for inside diameter measurement.
  • Includes 6 telescoping gages.
  • Features rounded tips to compensate for the radius.
  • Measures diameters from 8 mm to 150 mm.
  • Constructed from durable steel.
  • Minimum overall length is 1 "