Learn More
MSC Mitutoyo Thread Pitch Dia Measuring Wire, 40 TPI, 0.03 in Pitch
Catalog No. 5000513588
Encompass
The Mitutoyo thread pitch diameter measuring wire is engineered for accurate measurement of external thread pitch diameters. Featuring a 40 TPI (threads per inch) and a 0.03 in pitch, these wires are ideal for ensuring precision in manufacturing and quality control applications, particularly when used with thread plug gages.
  • Provides precise measurement of thread pitch diameters.
  • Features 40 threads per inch (TPI).
  • Equipped with a 0.03 in thread pitch.
  • Includes a wire length of 1.13 in.
  • Designed for use with thread plug gages.
  • Supplied as a 3-piece set for comprehensive measurements.
Catalog No. 50-005-13588 Supplier MSC Supplier No. 09182700
The Mitutoyo thread pitch diameter measuring wire is engineered for accurate measurement of external thread pitch diameters. Featuring a 40 TPI (threads per inch) and a 0.03 in pitch, these wires are ideal for ensuring precision in manufacturing and quality control applications, particularly when used with thread plug gages.
  • Provides precise measurement of thread pitch diameters.
  • Features 40 threads per inch (TPI).
  • Equipped with a 0.03 in thread pitch.
  • Includes a wire length of 1.13 in.
  • Designed for use with thread plug gages.
  • Supplied as a 3-piece set for comprehensive measurements.