Learn More
MSC Mitutoyo Thread Wire Set, 36 TPI, 0.01604 in Wire Diameter
Catalog No. 5000536183
Encompass
This 3-wire holder and thread wire set offers a fast and accurate method for thread measurement. It is compatible with micrometers featuring a 0.250 inch diameter anvil/spindle, ideal for monitoring pitch diameter variation and reducing measurement time.
  • Measures 36 threads per "
  • Features a wire diameter of 0.01604 "
  • Ensures fast and accurate thread measurement.
  • Compatible with micrometers having a 0.250 inch anvil/spindle.
  • Wires maintain a diameter tolerance of +/- 0.000010 "
  • Wires are lapped to a 2 micro inches AA surface finish and HRC 59-64 hardness.
  • Includes certification of accuracy, traceable to NIST.
  • Meets or exceeds all ANSI and ISO specifications.
Catalog No. 50-005-36183 Supplier MSC Supplier No. 09182718
This 3-wire holder and thread wire set offers a fast and accurate method for thread measurement. It is compatible with micrometers featuring a 0.250 inch diameter anvil/spindle, ideal for monitoring pitch diameter variation and reducing measurement time.
  • Measures 36 threads per "
  • Features a wire diameter of 0.01604 "
  • Ensures fast and accurate thread measurement.
  • Compatible with micrometers having a 0.250 inch anvil/spindle.
  • Wires maintain a diameter tolerance of +/- 0.000010 "
  • Wires are lapped to a 2 micro inches AA surface finish and HRC 59-64 hardness.
  • Includes certification of accuracy, traceable to NIST.
  • Meets or exceeds all ANSI and ISO specifications.