Learn More
MSC Mitutoyo Thread Wires, 3-Wire Holders, 22 TPI, NIST Traceable
Catalog No. 5000516764
Encompass
This 3-wire holder thread wire set offers a fast and accurate method for measuring thread plug gages. It effectively monitors wear and controls pitch diameter variation during thread fabrication, significantly reducing measurement time. Compatible with micrometers having a 0.250 in anvil/spindle diameter, this set helps ensure precise thread size control by detecting out-of-roundness and taper. It is suitable for both pre-plating and post-plating applications, ensuring quality control to demanding specifications. Includes accuracy certification and is traceable to NIST, meeting or exceeding all ANSI and ISO standards.
  • Threads per inch: 22.
  • Thread measuring wire diameter: 0.02624 in.
  • Wire diameter tolerance: +/- 0.000010 in.
  • Wire roundness tolerance: +/- 0.000010 in.
  • Surface finish: 2 micro inches AA, lapped.
  • Hardness: HRC 59-64.
Catalog No. 50-005-16764 Supplier MSC Supplier No. 09182783
This 3-wire holder thread wire set offers a fast and accurate method for measuring thread plug gages. It effectively monitors wear and controls pitch diameter variation during thread fabrication, significantly reducing measurement time. Compatible with micrometers having a 0.250 in anvil/spindle diameter, this set helps ensure precise thread size control by detecting out-of-roundness and taper. It is suitable for both pre-plating and post-plating applications, ensuring quality control to demanding specifications. Includes accuracy certification and is traceable to NIST, meeting or exceeding all ANSI and ISO standards.
  • Threads per inch: 22.
  • Thread measuring wire diameter: 0.02624 in.
  • Wire diameter tolerance: +/- 0.000010 in.
  • Wire roundness tolerance: +/- 0.000010 in.
  • Surface finish: 2 micro inches AA, lapped.
  • Hardness: HRC 59-64.