Learn More
MSC Mitutoyo U-Wave Fit Connection Unit, IP67 Caliper, IP65 Micrometer Compatible
Catalog No. 5000530735
Encompass
The U-Wave Fit Connection Unit is a wireless data transmitter designed to enhance the ergonomics and functionality of hand-held measuring instruments. This unit fits snugly behind the display, replacing traditional connecting cables with a rigid connector, thereby streamlining data transfer for various applications.
  • Fits to the back of hand-held measuring instruments.
  • Snug fit behind the display for an ergonomic feel, replacing connecting cables.
  • Approximately 20 m wireless transmission range.
  • Secure 2.4GHz wireless data transmission.
  • Easy data export to Excel or SPC software.
  • Data transfer confirmed by LED signal and optional buzzer.
  • IP67 transmitter ensures a coolant-proof gaging system.
  • Low power consumption, with 400,000 data transmissions per battery.
  • Compatible with IP67 calipers and IP65 coolant-proof micrometers.
Catalog No. 50-005-30735 Supplier MSC Supplier No. 90412362
The U-Wave Fit Connection Unit is a wireless data transmitter designed to enhance the ergonomics and functionality of hand-held measuring instruments. This unit fits snugly behind the display, replacing traditional connecting cables with a rigid connector, thereby streamlining data transfer for various applications.
  • Fits to the back of hand-held measuring instruments.
  • Snug fit behind the display for an ergonomic feel, replacing connecting cables.
  • Approximately 20 m wireless transmission range.
  • Secure 2.4GHz wireless data transmission.
  • Easy data export to Excel or SPC software.
  • Data transfer confirmed by LED signal and optional buzzer.
  • IP67 transmitter ensures a coolant-proof gaging system.
  • Low power consumption, with 400,000 data transmissions per battery.
  • Compatible with IP67 calipers and IP65 coolant-proof micrometers.