Learn More
MSC Mitutoyo Vernier Caliper, 0-130 mm / 0-5 inch, Fine Adjustment
Catalog No. 5000512444
Encompass
This Mitutoyo Series 532 Vernier Caliper is designed for precise measurements, offering both metric and inch scales. It features a fine-adjustment carriage for accurate slider movement, making it suitable for various measurement applications.
  • Fine-adjustment carriage for precise slider control
  • Measures outside diameter, inside diameter, depth, and steps
  • Includes a vinyl holster for protection
Catalog No. 50-005-12444 Supplier MSC Supplier No. 40086274
This Mitutoyo Series 532 Vernier Caliper is designed for precise measurements, offering both metric and inch scales. It features a fine-adjustment carriage for accurate slider movement, making it suitable for various measurement applications.
  • Fine-adjustment carriage for precise slider control
  • Measures outside diameter, inside diameter, depth, and steps
  • Includes a vinyl holster for protection