Learn More
MSC Mitutoyo Vertical Dial Test Indicator, 0.03 in Range, 0.0005 in Graduation, 0-15-0 Dial
Catalog No. 5000516894
Encompass
This dial test indicator is designed for precision measurements in narrow or recessed areas. It features a reversible measuring direction and an adjustable, carbide-tipped contact point for high accuracy. The one-piece movement ensures high-impact resistance and durability, while jeweled bearings provide high sensitivity and quick response. This vertical-type indicator is well-suited for centering holes under a machine tool spindle.
  • Easy-to-read dial graduations
  • Automatic reversal of measuring direction
  • Dust and oil protection with O-ring seal
  • Glare-free, scratch-resistant crystal
  • High-impact resistance and durability
  • High sensitivity and quick response
  • Supplied in a fitted plastic case
Catalog No. 50-005-16894 Supplier MSC Supplier No. 06250013
This dial test indicator is designed for precision measurements in narrow or recessed areas. It features a reversible measuring direction and an adjustable, carbide-tipped contact point for high accuracy. The one-piece movement ensures high-impact resistance and durability, while jeweled bearings provide high sensitivity and quick response. This vertical-type indicator is well-suited for centering holes under a machine tool spindle.
  • Easy-to-read dial graduations
  • Automatic reversal of measuring direction
  • Dust and oil protection with O-ring seal
  • Glare-free, scratch-resistant crystal
  • High-impact resistance and durability
  • High sensitivity and quick response
  • Supplied in a fitted plastic case