Learn More
MSC Mitutoyo Vertical Electronic Linear Scale, 0-8 in, 0.001 in Accuracy
Catalog No. 5000513534
Encompass
This vertical electronic linear scale is an absolute IP66 rated Digimatic scale designed for precise measurements. It offers a measurement range of 0 to 8 inches with 0.001 inch accuracy, and features inch/metric conversion, zero set, and data hold/output. Its robust design includes a hardened stainless steel blade and provides SPC data output for quality control applications.
  • Absolute IP66 Digimatic scale.
  • Vertical, single function scale.
  • Waterproof and dustproof protection (IP66).
  • Inch/metric conversion.
  • Zero set and data hold/output functions.
  • ABS Digimatic scale unit shows true position when turned on.
  • ORIGIN key allows setting of an absolute zero point.
  • Scale unit embedded in hardened stainless steel blade.
  • Includes two mounting brackets.
  • SPC data output.
  • Comes with one SR44 battery.
Catalog No. 50-005-13534 Supplier MSC Supplier No. 01129725
This vertical electronic linear scale is an absolute IP66 rated Digimatic scale designed for precise measurements. It offers a measurement range of 0 to 8 inches with 0.001 inch accuracy, and features inch/metric conversion, zero set, and data hold/output. Its robust design includes a hardened stainless steel blade and provides SPC data output for quality control applications.
  • Absolute IP66 Digimatic scale.
  • Vertical, single function scale.
  • Waterproof and dustproof protection (IP66).
  • Inch/metric conversion.
  • Zero set and data hold/output functions.
  • ABS Digimatic scale unit shows true position when turned on.
  • ORIGIN key allows setting of an absolute zero point.
  • Scale unit embedded in hardened stainless steel blade.
  • Includes two mounting brackets.
  • SPC data output.
  • Comes with one SR44 battery.