Learn More
Medchemexpress LLC Mivacurium dichloride | 106861-44-3 | 99.0% | 25 MG
SDP
Catalog No. 5000414108
Encompass_Preferred
Mivacurium dichloride is a benzylisoquinoline derivative and a short-acting non-depolarizing neuromuscular blocking agent and skeletal muscle relaxant. It functions by coupling with the nAChR to reduce or inhibit the depolarizing effect of acetylcholine on the muscle cell's terminal disc.
  • Short-acting non-depolarizing neuromuscular blocking agent.
  • Skeletal muscle relaxant.
  • Functions by inhibiting acetylcholine effects on muscle cells.
  • Induces LAD2 cell degranulation in a dose-dependent manner.
  • Rapid hydrolysis in plasma.
  • Short duration of action (less than 10 minutes).
  • Non-neurological toxicity.
  • Does not alter heart rate.
Catalog No. 50-004-14108 Supplier Medchemexpress LLC Supplier No. 25MGHYB1700A
Mivacurium dichloride is a benzylisoquinoline derivative and a short-acting non-depolarizing neuromuscular blocking agent and skeletal muscle relaxant. It functions by coupling with the nAChR to reduce or inhibit the depolarizing effect of acetylcholine on the muscle cell's terminal disc.
  • Short-acting non-depolarizing neuromuscular blocking agent.
  • Skeletal muscle relaxant.
  • Functions by inhibiting acetylcholine effects on muscle cells.
  • Induces LAD2 cell degranulation in a dose-dependent manner.
  • Rapid hydrolysis in plasma.
  • Short duration of action (less than 10 minutes).
  • Non-neurological toxicity.
  • Does not alter heart rate.