Learn More
TARGETMOL CHEMICALS INC Mivacurium dichloride 25MG
SDP
Catalog No. 502877690
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Mivacurium dichloride (Mivacurium chloride) is a non-depolarising, rapid neuromuscular blocker of short duration, used therefore as anesthetic, to provide skeletal muscle relaxation in minor surgical operations, in emergency surgical procedures of short to intermediate duration and during intubation of the trachea. Purity 99.52%
Catalog No. 50-287-7690 Supplier TARGETMOL CHEMICALS INC Supplier No. T2237125MG
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Mivacurium dichloride (Mivacurium chloride) is a non-depolarising, rapid neuromuscular blocker of short duration, used therefore as anesthetic, to provide skeletal muscle relaxation in minor surgical operations, in emergency surgical procedures of short to intermediate duration and during intubation of the trachea. Purity 99.52%