Learn More
Medchemexpress LLC Mivavotinib | 1312691-33-0 | 344.39 g/mol | C17H21FN6O | 50 MG
SDP
Catalog No. 5000212914
Encompass_Preferred
Mivavotinib is a small-molecule research inhibitor with dual activity against spleen tyrosine kinase (SYK) and FMS-like tyrosine kinase 3 (FLT3). Supplied as a laboratory-grade powder, it is intended for research and analytical use only and is not for human therapeutic applications. The compound is characterized by CAS number 1312691-33-0 and a molecular weight of approximately 344.39 g/mol.
  • dual inhibition of SYK and FLT3 for mechanistic studies
  • supplied as a stable powder suitable for analytical workflows
  • well-characterized by CAS and molecular-weight identifiers
  • available in small research-friendly package sizes
  • intended for research use with appropriate safety precautions
Catalog No. 50-002-12914 Supplier Medchemexpress LLC Supplier No. HY100867A50MG
Mivavotinib is a small-molecule research inhibitor with dual activity against spleen tyrosine kinase (SYK) and FMS-like tyrosine kinase 3 (FLT3). Supplied as a laboratory-grade powder, it is intended for research and analytical use only and is not for human therapeutic applications. The compound is characterized by CAS number 1312691-33-0 and a molecular weight of approximately 344.39 g/mol.
  • dual inhibition of SYK and FLT3 for mechanistic studies
  • supplied as a stable powder suitable for analytical workflows
  • well-characterized by CAS and molecular-weight identifiers
  • available in small research-friendly package sizes
  • intended for research use with appropriate safety precautions