Learn More
Medchemexpress LLC Mivavotinib monohydrochloride | 1312691-33-0 | 344.39 | C17H21FN6O | 25 MG
SDP
Catalog No. 5000212355
Encompass_Preferred
Mivavotinib monohydrochloride is a research-grade small-molecule inhibitor of spleen tyrosine kinase (SYK) and fms-related tyrosine kinase 3 (FLT3), used to investigate kinase signaling and antitumor activity in preclinical studies. The compound is provided as a solid for formulation and assay use.
  • Potent dual SYK and FLT3 inhibition with low-nanomolar activity.
  • Suitable for biochemical and cell-based assays.
  • Useful as a tool compound in oncology and signal-transduction research.
  • Molecular formula C17H21FN6O and molecular weight 344.39 g/mol.
Catalog No. 50-002-12355 Supplier Medchemexpress LLC Supplier No. HY100867A25MG
Mivavotinib monohydrochloride is a research-grade small-molecule inhibitor of spleen tyrosine kinase (SYK) and fms-related tyrosine kinase 3 (FLT3), used to investigate kinase signaling and antitumor activity in preclinical studies. The compound is provided as a solid for formulation and assay use.
  • Potent dual SYK and FLT3 inhibition with low-nanomolar activity.
  • Suitable for biochemical and cell-based assays.
  • Useful as a tool compound in oncology and signal-transduction research.
  • Molecular formula C17H21FN6O and molecular weight 344.39 g/mol.